Learn More
IFN-gamma R1/CD119 Mouse anti-Human, Alexa Fluor 350, Clone: 92101, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB6731U-100UG
This item is not returnable.
View return policy
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Flow Cytometry.Specifications
| IFN-gamma R1/CD119 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P15260.1 | |
| IFNGR1 | |
| Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R1 Glu18-Gly245 Accession # P15260.1 | |
| RUO | |
| 3459 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 92101 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IFN-gamma R1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta . | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
Spot an opportunity for improvement?
For Research Use Only (RUO)