Learn More
Fisherbrand™ Grade 12 Ashless Medium-Fast Filtering Quantitative Filter Paper
Shop All Fisherbrand ProductsDescription
- Made with 100% cotton linter fibers
- Washed with acid and cleaned with demineralized water to obtain high purity
- Ash content is 0.007%
- Applications: Chemical, food, beverage industries and atmospheric analysis
Specifications
Specifications
| Description | Ash Content: 0.007% |
| Filter Type | Ashless |
| Format | Disc |
| Material | Cotton Linters |
| Particle Retention | 8 to 12 μm |
| Pore Size | 8 to 12 μm |
| Filtration Speed | 20 sec. |
| For Use With (Application) | For coarser precipitates such as lead- iron- and silver sulphide, alkali carbonates, lead chromate, arsenic, antimony, cadmium, food stuffs |
| Grade | 12 |
| Quantity | 100/Pk. |
| Show More |