Learn More
Heidolph™ PharMed™ Tubing
Product Code. 10584542 Shop All Heidolph Instruments Products
Click to view available options
Tubing Size:
0.8 mm I.D./4.0 mm O.D.
1.7 mm I.D/4.9 mm O.D.
3.1 mm I.D./6.3 mm O.D.
4.8 mm I.D./8.0 mm O.D.
6.4 mm I.D./11.3 mm O.D
Unit Size:
Each
Product Code. 10584542
Brand: Heidolph™ 5252900000
Only null left
Add to basket
Add to basket
Ideal tubing for medical, laboratory, and research uses. Heidolph™ PharMed™ Tubing is durable and non-toxic.
  • High fatigue strength under repeated reversed bending stresses.
  • Non-toxic, biocompatible.
  • Very low gas permeability.
  • Well suited to acids and bases.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance FDA (21 CFR 177.2600), USP Class VI, GLP, Pharmacopoea and Europaea
Autoclavable Autoclavable
Diameter (Metric) Inner 6.4 mm
Material Thermoplastic
Diameter (Metric) Outer 11.3 mm
Thickness (Metric) 2.5 mm
For Use With Pump
For Use With (Application) High Fatigue Strength, Non-Toxic, Biocompatible, Very Low Gas Permeability, Well-Suited to Acids and Bases.
Min. Operating Temperature (Metric) 40°C
Sterility Non-sterile
Tubing Size 6.4 mm I.D./11.3 mm O.D
Sterilization Method Autoclave, Ethylene Oxide, Irradiation
Height (Metric) 8.8 m Water
Gas Permeability Very low
Limitations Additives may migrate.
Maximum Pressure Continuous/short time 0.8/1.8 bar
Vacuum Level Excellent
Max. Operating Temperature (Metric) 75°C
Product Type Pump Tubing