Description
- PVC
- Shallow form
- Thick sides, shatterproof
- Largely resistant to acids and alkalis
- Temperature resistant from -30°C to +70°C
Specifications
Specifications
| Dimensions (L x W) | 310 x 410 mm Inner Base, 370 x 470 mm Outer Rim |
| Color | Red |
| Height (Metric) | 60 mm |
| Length (Metric) | 310 mm (Inner Base), 370 mm (Outer Base) |
| Material | PVC |
| Type | Photographic Tray |
| Width (Metric) | 410 mm (Inner Base), 470 mm (Outer Base) |
| Height (English) | 2.36 in. |
| Shape | Rectangular |