Learn More
Macherey-Nagel™ Adamant™ Unmodified Standard Silica Layers for TLC, UV254
Product Code. 10101762
Change view
Click to view available options
Dimensions (L x W):
10 x 10 cm
2.5 x 7.5 cm
5 x 10 cm
5 x 20 cm
Quantity:
100 Pk.
200 Pk.
25 Pk.
Unit Size:
Pack of 100
Pack of 200
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity unitSize
10101762 5 x 20 cm 100 Pk. Pack of 100
10416352 2.5 x 7.5 cm 100 Pk. Pack of 100
10779584 5 x 10 cm 200 Pk. Pack of 200
10070542 10 x 10 cm 25 Pk. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10101762 Supplier Macherey-Nagel™ Supplier No. 821015
Only null left
Add to basket
Add to basket
Increase separation efficiency and maximize resolution during TLC (thin-layer chromatography). Adamant™ Unmodified Standard Silica Layers for TLC, UV254, are available on glass plates with an optimized binder system and UV indicator.
  • Adamant™ plates feature a newly developed binder system, which results in an outstanding hardness of the silica layer
  • The plate can be easily labelled for clear identification
  • The abrasion resistance prevents undesired changes or damage to the layer during transportation or handling
  • A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber
  • With fluorescent indicator at 254nm
  • 250μm layer thicknessSilica 60, specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g, 
  • Particle size 5–17 μm
  • Increased separation efficiency due to an optimized particle size distribution
  • High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer
  • Available as glass plates 
Specifications
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 100 Pk.
Type Glass Plate
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.25 mm
Dimensions (L x W) 5 x 20 cm