Learn More
RUBIS Full Plastic Tweezers, ESD Safe
Click to view available options
Tip Style:
Angled, Pointed, Ultra Fine
Round, Flat
Squared, Fine
Straight, Round, Flat
Unit Size:
Each
Description
- Vestamid Plastic
- ESD Safe
- Avoids scratches on delicate components
- Very good mechanical resistance
- Easy to use
- High Precision
- 100% Made in Switzerland
Specifications
Specifications
| Tip Style | Squared, Fine |
| Length (English) | 4.72 in. |
| Material | ESD Safe Vestamid Plastic |
| Length (Metric) | 120 mm |
| Product Type | Plastic Tweezers |
| Quantity (No. per Case) | Each |
| Type | Tweezers |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.