Learn More
OHAUS™ Vessel Holder for Microplate Vortex Mixer
Shop All Ohaus Products
Click to view available options
Holds:
125 and 250 mL Erlenmeyer Flasks
500 and 1000 mL Erlenmeyer Flasks
Unit Size:
Each
Description
- For use with Ohaus Microplate Vortex Mixer
Specifications
Specifications
| Holds | 500 and 1000 mL Erlenmeyer Flasks |
| Product Type | Large Vessel Holder |
| Material | Plasitc |
| Description | Large Vessel Holder For 500 ml/1 L Flask |
| For Use With (Equipment) | Vortex Mixer |
| Includes | Grip mat |
| Packaging Quantity | 1 Ea. |