Learn More
Scientific Industries SI™ Vertical Microtube Holder
Product Code. 15517375 Shop All Ohaus Products
Change view
Click to view available options
No. of Positions:
9
24
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
15517375 24 Each
15587365 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15517375 Supplier Scientific Industries SI™ Supplier No. 83041240
  • ONLINE EXCLUSIVE PRICE
    145.00€
    127.43€ / Pack of 1
    Save 17.57€
    12% Off
In Stock
Requires no additional attachment for use. Scientific Industries SI™ Vertical Microtube Holder snaps microtubes into place.
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vortex-Genie 2 Mixer and Vortex-Genie Pulse
No. of Positions 24