Learn More
Texwipe™ Vertex™ Sterile High Sorption Wipers
Product Code. 16385488 Shop All Texwipe Products
Click to view available options
Length (English):
9 in.
12 in.
Description:
Dry
Pre-wetted
Unit Size:
Pack of 125
Pack of 20
Pack of 25
Pack of 500
Product Code. 16385488
Brand: Texwipe™ TX3042P
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
100% continuous-filament, polyester knit fabric wipers made in the USA from virgin stock
Sealed-edge sterile wipers, dry or pre-wetted
  • Soft feel with high sorption capacity
  • Confirm radiation dose met specified dose at time of receiving
  • Processed on Texwipe's Vertex™ manufacturing system
  • Quarterly microbial testing by Agarose Overlay and Limulus Amebocyte Lysate (LAL) endotoxin testing
Pre-Wetted Wipers
  • 70% USP-Grade IPA/30% USP Purified Water
Order Info
Triple-bagged
Specifications
Product Type High Sorption Wiper
Length (English) 12 in.
Length (Metric) 30.5 cm
Width (English) 12 in.
Width (Metric) 30.5 cm
Material Polyester
Certifications/Compliance ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Description Pre-wetted
Edges Sealed
Dimensions (L x W) 12 x 12 in. (30.48 x 30.48 cm)
For Use With (Application) Ideal for cleaning delicate surfaces or applications
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Texwipe Wipers pre-wetted with 60% - 70 % IPA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : isopropanol