Learn More
Thermo Scientific™ Versette™ Automated Liquid Handler
Product Code. p-4528664 Shop All Thermo Scientific Products
Click to view available options
Description:
6-position stage for Versette, guarding included
96- and 384- channel pipetting module for use with 96- and 384-channel Versette pipetting heads
Versette Base Unit Only
Versette instrument (includes base, pipetting module and a 6-position stage)
Versette system for MSIA application (includes base, pipetting module, 6-position stage, 96-channel pipetting head for volumes of 5-300μl and a serial dilute magazine 96/300μl)
Unit Size:
Each
Product Code. 11684912
Brand: Thermo Scientific™ 65001BS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Support a wide range of applications with this compact handler, featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads.
Support a wide range of applications with the Thermo Scientific™ Versette™ Automated Liquid Handler, a compact handler featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads. An ideal choice for the liquid handling needs of most laboratories, it supports 96/384 plate replication, plate stamping and serial dilution procedures.
Support a wide range of applications, including high-throughput Mass Spectrometric Immunoassay (MSIA), plate stamping, plate reformatting, serial dilution and many other applications.
96-and 384-channel pipetting heads:
  • Total volume range 0.5-300μL
  • All pipetting heads include RFID tags for self-identification
Consumables for optimized performance:
  • All pipetting heads utilize Thermo Scientific™ D.A.R.T.'S
Compact size:
  • Six position stage uses a unique dual-level design to minimize space requirements
  • Fits on standard bench and biological hoods
User-friendly programming options:
  • Simple pipetting procedures can be created using visual on-board graphical display
  • For more complex protocols and advanced editing, Thermo Scientific™ ControlMate™ software is available
TRUSTED_SUSTAINABILITY
Specifications
Product Type Versette Base Unit
Description Versette Base Unit Only
Channels 96, 384
Volume (Metric) 0.5 to 300 μL
For Use With (Equipment) 96-, 384- well plates with different heights, storage tubes in rack (Thermo Scientific 0.5 to 1.4mL
Interface RS-232
Operating System Windows XP, Windows Vista (32-and 64-bit), Windows 7 (32- and 64-bit)
Width (English) 27 in.
Width (Metric) 68 cm
Height (English) 27 in.
Height (Metric) 68 cm
Weight (English) 80 lb.
Weight (Metric) 36 kg
Electrical Requirements 100 to 240V 50/60Hz
Voltage 100/240 V
Frequency 50/60 Hz