missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VersaTREK™ Myco Growth Supplement (Myco GS)

Product Code. 15349231
Change view
Click to view available options
Packaging Type:
50 mL Each
Media Type:
Myco growth supplement
Quantity:
50 mL
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Type Media Type Quantity Unit Size
15349231 50 mL Each Myco growth supplement 50 mL Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15349231 Supplier Thermo Scientific™ Supplier No. 711242

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Promote growth of slow-growing mycobacteria within an inoculated bottle and facilitate detection with Thermo Scientific™ VersaTREK™ Myco Growth Supplement (Myco GS).

Promote growth of slow-growing mycobacteria within an inoculated bottle for enhanced detection with Thermo Scientific™ VersaTREK™ Myco Growth Supplement (Myco GS).

The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.

Not all products are available for sale in all territories. Please inquire.

VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Must be stored in the dark
Description VersaTREK Myco Growth Supplement (Myco GS)
Media Type Myco growth supplement
Product Type Microbiology Media
Target Organism Class Mycobacteria
Test Type Microbial Detection, Antimicrobial Susceptibility
No. per Pack 5
Packaging Type 50 mL Each
Quantity 50 mL
Specimen Type All Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
Species Mycobacterium tuberculosis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.