Learn More
Thermo Scientific™ VersaLid for VersaCool™ Refrigerated Circulating Bath, reversible hinged lid
Shop All Thermo Scientific ProductsDescription
Utilize the VersaLid for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Configurable to the user’s comfort, the VersaLid offers a tool-less setting for left or right-handed hinge movement and also for no hinge movement. It’s dripless when used in hinge mode.
- One lid that can be left-handed hinge, right-handed hinge, or unhinged lid
- Drip-less lid reduces moisture and directs condensate back into bath
Specifications
Specifications
| Description | VersaLid-reversible hinged lid |
| For Use With (Equipment) | Thermo Scientific VersaCool Refrigerated Circulating Bath |
| Type | Cover |