Learn More
3M™ Versaflo™ Powered Air Respirator System Healthcare Ready Kit, TR-300E+ HKL
Product Code. 17887593 Shop All 3M Products
Change view
Click to view available options
Product Type:
PAPR Healthcare Systems and Kits
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17887593 PAPR Healthcare Systems and Kits Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17887593 Supplier 3M™ Supplier No. 7100233765
  • ONLINE EXCLUSIVE PRICE
    1865.00€
    1215.64€ / Pack of 1
    Save 649.36€
    34% Off
 Restricted Item
Estimated Shipment: 04-01-2027
3M™ Versaflo™ TR-300+ Powered Air Respirators are versatile and easy-to-use respiratory protection against particulates hazards.
  • Slim, lightweight unit is well-balanced and comfortable to wear for long work shifts
  • Two user selectable airflows enable the user to control flow settings to allow the user to find the setting they find most comfortable
  • Powered Air Respirators automatically adjust airflow to compensate for filter loading to deliver consistent filtered air during use
  • Audible and visual alerts for low airflow and low battery charge-level to keep you prepared and informed
  • Able to be worn on a belt or backpack
  • Available in convienient, easy to buy assemblies and kits
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 9 to 12 hr.
Battery Type High Capacity Lithium Ion - Rechargeable
Cartridge/Filter PARTICULATE
Color Black
Product Type PAPR Healthcare Systems and Kits
Size One Size
For Use With (Equipment) TR-3712E Filter
Includes S-133L Headcover
Intrinsic Safety (IS) Rating No
Model TR-300E+ HKL