Learn More
3M™ Versaflo™ TR-3712E P3 Particulate Filter
Description
- High efficiency P3 particulate filter
- Quick and easy changeover by hand
- EN 12941, CE Approved
- For use with the 3M™ TR-300+ Powered Air Respirator series
Specifications
Specifications
| Certifications/Compliance | EN 12941, CE Approved |
| Filter Type | P3 |
| Product Type | Particulates Filter |
| For Use With (Equipment) | Powered Air Respirator TR-300+. |