Learn More
3M™ Versaflo™ Battery Attachment Tool TR-838/94244(AAD)
Product Code. 16436360 Shop All 3M Products
Change view
Click to view available options
For Use With: TR-802E Powered Air Turbo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16436360 For Use With: TR-802E Powered Air Turbo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16436360 Supplier 3M™ Supplier No. 7100150921
  • ONLINE EXCLUSIVE PRICE
    25.00€
    19.49€ / Pack of 1
    Save 5.51€
    22% Off
Estimated Shipment: 16-10-2026
3M™ Versaflo™ Battery Attachment Tool TR-838/94244(AAD) is an accessory for attaching TR-830 batteries to the TR-802E Powered Air Turbo.
  • Enables attachment of TR-830 battery to TR-802E Powered Air Turbo
  • Ensures secure fastening
  • Prevents accidental battery removal and spark risk during use
  • The mechanical fastening action prevents accidental battery removal and reduces spark risk during use per EN 60079-11
  • Intrinsically safe and suitable for potentially explosive atmospheres
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, NIOSH Approved
Product Type Battery Attachment Tool
For Use With (Equipment) TR-802E Powered Air Turbo