Description
- Adaptor helps fit the turbo unit to the backpack harness, so it can be worn over the shoulders
- Easy to connect to turbo unit
- For use with the 3M™ Versaflo™ BPK-01 backpack
Specifications
Specifications
| Color | Black |
| Product Type | Backpack Adapter |
| For Use With (Equipment) | 3M™ Versaflo™ Powered Air Turbo TR-600 and BPK-01 Backpack |