Learn More
Cole-Parmer™ Rapid Surface Hygiene Kit
Product Code. 11740559 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11740559 50 Tests Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11740559 Supplier Cole-Parmer™ Supplier No. 1404030
  • ONLINE EXCLUSIVE PRICE
    214.00€
    176.34€ / Pack of 50
    Save 37.66€
    17% Off
Estimated Shipment: 17-12-2026
Use this Rapid Surface Hygiene Kit to detect invisible food residue without an instrument-in only 10 to 60 seconds.
  • VERIcleen is sensitive to food residues including meat, dairy, bakery, beverages, eggs, fish, etc. It is an inexpensive rapid hygiene program to the foodservice, retail and institutional industries.
  • Applications include HACCP programs, hand washing hygiene compliance, and overall sanitation monitoring.
  • Application is extremely simple, just spray to wet-dry surface and swab.
  • A ''clean surface'' shows no purple until after 3 minutes.
  • A ''Not clean'' surface shows purple in 10 to 60 seconds.
  • Each kit includes 50 tests in a light-tight snap-top container, 1 bottle of wetting liquid with an atomizer, instruction leaflet, and desiccant packet.
    • If desiccant inside the packet has turned from blue to pink, the strips have been stored improperly, and may be invalid.
Specifications
Description Rapid Surface Hygiene Kit, 50 Tests/Kit
Quantity 50 Tests
Sample Type Environmental Sample, Food Sample