Learn More
Phenomenex™ Verex™ Crimp Seals, 11 mm, with Septa, No Slit
Description
- Analysis: Cleanliness and performance tested as standard
- Certification: 100% defect-free
- Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
Specifications
Specifications
| Color | Silver |
| Diameter (Metric) Outer | 11 mm |
| Pre-slit | No |
| Sterility | Sterile |
| Septum | PTFE/Rubber |
| Quantity | 100 Pk. |
| Closure Size | 11 mm |
| Marking Spot | No |
| Autoclavable | Autoclavable |
| Product Type | Cap |