Learn More
Emma Safety Footwear Vera Safety Shoes
Product Code. 15998551
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
46
47
48
49
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998551 38 1 pair
15968551 35 1 pair
15978551 36 1 pair
15988551 37 1 pair
15908561 39 1 pair
15918561 40 1 pair
15928561 41 1 pair
15938561 42 1 pair
15948561 43 1 pair
15958561 44 1 pair
15978561 46 1 pair
15988561 47 1 pair
15998561 48 1 pair
15908571 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15998551 Supplier Emma Safety Footwear Supplier No. MM50654438
  • ONLINE EXCLUSIVE PRICE
    139.65€
    73.50€ / 1 pairs
    Save 66.15€
    47% Off
View all promotions
Estimated Shipment: 17-12-2026
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and direct resistant.
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Elastic
Color White
Design Vera
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 38
Toe Material Steel