Läs mer
Emma Safety Footwear Vera Safety Shoes
Produktkod. 15948561
Change view
Klicka för att se tillgängliga alternativ
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
Förpackningsstorlek:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Size unitSize
15948561 43 1 pair
15968551 35 1 pair
15978551 36 1 pair
15988551 37 1 pair
15998551 38 1 pair
15908561 39 1 pair
15918561 40 1 pair
15928561 41 1 pair
15938561 42 1 pair
15958561 44 1 pair
15968561 45 1 pair
15978561 46 1 pair
15988561 47 1 pair
15998561 48 1 pair
15908571 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15948561 Supplier Emma Safety Footwear Supplier No. MM50654443
  • ONLINE EXCLUSIVE PRICE
    127.05€
    73.50€ / 1 pairs
    Save 53.55€
    42% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and direct resistant.
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Elastic
Color White
Design Vera
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 43
Toe Material Steel