Learn More
Velpatasvir, TRC
Specifications
Specifications
| Chemical Name or Material | Velpatasvir |
| CAS | 1377049-84-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C49H54N8O8 |
| Quantity | 100 mg |
| Synonym | GS 5816, Methyl [(2S)-1-[(2S,5S)-2-[9-[2-[(2S,4S)-1-[(2R)-2-[(Methoxycarbonyl)amino]-2-phenylacetyl]-4-(methoxymethyl)pyrrolidin-2-yl]-1H-imidazol-5-yl]-1,11-dihydroisochromeno[4',3':6,7]naphtho[1,2-d]imidazol-2-yl]-5-methylpyrrolidin-1-yl]-3-methyl-1-oxobutan-2-yl]carbamate |
| InChI Formula | InChI=1S/C49H54N8O8/c1-26(2)41(54-48(60)63-5)47(59)57-27(3)12-17-38(57)45-51-36-16-14-30-20-35-33-15-13-31(19-32(33)25-65-40(35)21-34(30)43(36)53-45)37-22-50-44(52-37)39-18-28(24-62-4)23-56(39)46(58)42(55-49(61)64-6)29-10-8-7-9-11-29/h7-11,13-16,19-22,26-28,38-39,41-42H,12,17-18,23-25H2,1-6H3,(H,50,52)(H,51,53)(H,54,60)(H,55,61)/t27-,28-,38-,39-,41-,42+/m0/s1 |
| SMILES | CC([C@H](NC(OC)=O)C(N1[C@H](CC[C@H]1C2=NC3=C(N2)C4=CC5=C(C6=C(CO5)C=C(C7=CN=C(N7)[C@@H]8C[C@H](COC)CN8C([C@H](NC(OC)=O)C9=CC=CC=C9)=O)C=C6)C=C4C=C3)C)=O)C |
| Molecular Weight (g/mol) | 883 |
| Formula Weight | 882.4065 |
| Show More |