Learn More
Novus Biologicals™ VECTASTAIN(R) ABC anti-mouse/rabbit AP Immunodetection Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Content And Storage | Store at 4°C. Do not freeze. |
| For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), ISH, ELISpot |
| Species | Mouse, Rabbit |
| Quantity | 1 Kit |