Learn More
Ekastu Safety™ Varnish and Welding Respirator
Description
- Protects against toxic and highly toxic particles as welding fume, paint and varnish particles (up to 50 times the NPF value) as well as solvents below Occupational Exposure Limit
- With Cool Down exhalation valve
- With color-coded nosepiece and exhalation valve
- Activated charcoal layer included
- Continous adjustable harness
- Broad cotton-lined inner sealing lip guarantees close and comfortable fit as well as safety
- Filter class DIN EN 149:2001 FFP3 NR
- Specially designed for wearer of EKASTU spectacles and goggles
- In original packaging: 5 years shelf life
- 5 pieces per pack
Specifications
Specifications
| Color | Red |
| Filter Type | FFP3 |
| Product Type | Welding Particulate Respirator |
| With Exhalation Valve | Yes |
| Includes | Color-coded nosepiece and exhalation valve; Activated charcoal layer |
| Quantity | 5 Pk. |
| Weight (Metric) | 27 g |