Learn More
WTW™ Variable Wave-length Thermoreactor with 4 Temperature Settings
Click to view available options
Holds:
12 Reaction Cuvettes
2 x 12 Reaction Cuvettes
2 x 12 Reaction Cuvettes in 2 heating blocks
Unit Size:
Each
Description
Standardized digestion programs for the standard parameters COD, quick COD (148°C, 20 min.), total nitrogen, total phosphorus, TOC and heavy metals of galvanizing with automatic heating and switch-off. 8 user-defined digestions at temperatures between 25-170°C can be programmed for special degradations and therefore also allow high temperature digestions. Two different programs for 12 cuvettes each can be run at the same time. You can use an external temperature sensor instead of a vial with data output to the PC/printer for analytical quality assurance (AQA).
Specifications
Specifications
| Product Type | Thermoreactor |
| Hertz | 50/60 Hz |
| Voltage | 230/115 V |
| Wattage | 560 W |
| Depth (Metric) | 315 mm |
| Electrical Requirements | 115/230 V ±15%, 50/60 Hz, 560 W |
| For Use With (Application) | Routine and specialized tasks in wastewater and in laboratories |
| Height (Metric) | 185 mm |
| Holds | 2 x 12 Reaction Cuvettes in 2 heating blocks |
| Width (Metric) | 256 mm |