Learn More
Vanillylmandelic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Vanillylmandelic Acid Ethyl Ester |
| CAS | 52058-11-4 |
| Molecular Formula | C11H14O5 |
| Quantity | 2.5 g |
| Synonym | 4-Hydroxy-3-methoxy-mandelic Acid Ethyl Ester, alpha,4-Dihydroxy-3-methoxy-benzeneacetic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C11H14O5/c1-3-16-11(14)10(13)7-4-5-8(12)9(6-7)15-2/h4-6,10,12-13H,3H2,1-2H3 |
| SMILES | CCOC(=O)C(O)c1ccc(O)c(OC)c1 |
| IUPAC Name | ethyl 2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetate |
| Molecular Weight (g/mol) | 226.23 |
| Formula Weight | 226.0841 |