Learn More
Vancomycin Monohydrochloride, TRC
Product Code. 30456942
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456942 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456942 Supplier Toronto Research Chemicals Supplier No. TRCV096500500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Vancomycin Monohydrochloride
CAS 197638-25-8
Molecular Formula C66H75Cl2N9O24 • HCl
Quantity 500 mg
Synonym Lyphocin Monohydrochloride, Meek Monohydrochloride, Vancocin Monohydrochloride, Vancocine Monohydrochloride, Vancocyn Monohydrochloride, Vancor Monohydrochloride
InChI Formula InChI=1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42?,44-,46?,47+,48-,49+,50-,51+,52+,53?,54?,56+,57+,65-,66-;/m0./s1
SMILES [H][C@@]([C@](NC([C@@](NC)([H])CC(C)C)=O)([H])C1=O)(O)C(C=C2Cl)=CC=C2OC3=C(O[C@@H]4OC(CO)C(O)C(O)[C@H]4O[C@@H]5O[C@@H](C)[C@@H](O)[C@](N)(C)C5)C(OC6=CC=C([C@]7(O)[H])C=C6Cl)=CC([C@]([H])(C(NC8([H])C(N[C@]7([H])C(N[C@]([H])(C(O)=O)C(C=C(O)C=C9O)=C9C%10=C(O)C=CC8=C%10)=O)=O)=O)NC([C@@](CC(N)=O)([H])N1)=O)=C3.Cl
Molecular Weight (g/mol) 1449.253646
Formula Weight 1483.4069