Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Product Code. 11947471 Shop All Thermo Scientific Products
Click to view available options
Type:
GENESYS Validator
Performance Verification Kit
Validator 2 for INSIGHT – A4
Validator Lite
Unit Size:
Each
Product Code. 11947471
Brand: Thermo Scientific™ 699119300
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Packages provide:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ), Operational Qualification (OQ) and Performance Qualification (PQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance
  • Installation and instrument certification
TRUSTED_SUSTAINABILITY
Specifications
Type Validator Lite
For Use With (Equipment) Evolution 60S/60; BioMate 3S; GENESYS 10S UV-Vis Bio/10S UV-Vis
Model UV Validator Lite