Learn More
Valbenazine (~90% Purity), TRC
Product Code. 30452450
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452450 25 mg 25mg
30456414 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30452450 Supplier Toronto Research Chemicals Supplier No. TRCV08700025MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1025504-45-3
Molecular Formula C24 H38 N2 O4
Molecular Weight (g/mol) 418.57
Synonym L-Valine, (2R,3R,11bR)-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-yl ester, Ingrezza, NBI 98854, Valbenazine, (2R,3R,11bR)-9,10-dimethoxy-3-(2-methylpropyl)-1,3,4,6,7,11b hexahydro-2H-pyrido[2,1-a]isoquinolin-2-yl L-valinate
IUPAC Name [(2R,3R,11bR)-9,10-dimethoxy-3-(2-methylpropyl)-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizin-2-yl] (2S)-2-amino-3-methylbutanoate
SMILES COc1cc2CCN3C[C@@H](CC(C)C)[C@@H](C[C@@H]3c2cc1OC)OC(=O)[C@@H](N)C(C)C
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C24H38N2O4/c1-14(2)9-17-13-26-8-7-16-10-21(28-5)22(29-6)11-18(16)19(26)12-20(17)30-24(27)23(25)15(3)4/h10-11,14-15,17,19-20,23H,7-9,12-13,25H2,1-6H3/t17-,19-,20-,23+/m1/s1
Formula Weight 418.2832
Chemical Name or Material Valbenazine