Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Straight
Product Code. p-7222565
Click to view available options
Plug Bore:
4 mm
6 mm
8 mm
10 mm
missing translation for 'unitSize'
Each
Product Code. 10090221
missing translation for 'mfr': Lenz Laborglasintrumente 02901804
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With straight side arms and hollow glass plug
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Plug Bore 4 mm
Material Borosilicate Glass (3.3)