Learn More
Lenz Laborglasintrumente Vacuum Stopcocks, 1-Way, Parallel
Click to view available options
Plug Bore:
2.5 mm
4 mm
6 mm
8 mm
10 mm
Unit Size:
Each
Description
- Acc. to DIN 12545 and in addition to DIN standard
- With parallel side arms and hollow glass plug
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
Specifications
| Plug Bore | 8 mm |
| Material | Borosilicate Glass (3.3) |