Learn More
KNF™ Diaphragm Vacuum Pump System SC 920 G
Click to view available options
Flow Rate:
1.26 m³/hr.
Unit Size:
Each
Description
- Automatic, accurate recognition and monitoring of the boiling point using the integrated ramp function.
- Speed-controlled.
- Integrated gas ballast valve.
Specifications
Specifications
| Max. Vacuum | 2 Mbar abs. |
| Dimensions (L x W x H) | 366 x 423 x 294 mm |
| Material | Pump head: PPS, Diaphragm: PTFE-coated, Valves: FFPM |
| Weight (Metric) | 15.2 kg |
| Temperature (Metric) Max | 40°C |
| Flow Rate | 1.26 m³/hr. |