Learn More
Fisherbrand™ Vacuum Pump Oil
Shop All Fisherbrand Products
Click to view available options
Quantity:
5 L
Unit Size:
Each
Description
Generic mineral oil for use in most one and two stage rotary vane pumps.
Specifications
Specifications
| Product Type | Vacuum Pump Oil |
| For Use With (Application) | For General Use in Rotary Vane Pumps |
| Quantity | 5 L |
| Volume (Metric) | 5 L |