Learn More
Memmert Vacuum pump 230V 50Hz with chemically resistant 4x diaphragm, app 50l/min, 3m3/h, for VO101
Brand:  Memmert VO101E07509
Vacuum pump 230V 50Hz with chemically resistant 4x diaphragm, app 50l/min, 3m3/h, for VO101
Product Code. 16328037
  • ONLINE EXCLUSIVE PRICE
    4720.00€
    4326.00€ / Each
    Save 394.00€
    8% Off
Only null left
Estimated Shipment: 12-03-2026