Learn More
BOLA Vacuum Adaptor with Ground Joint
Product Code. 15649418
Click to view available options
:
22 mm
Product Code. 15649418
Brand: BOLA N165506
Only null left
Add to basket
Add to basket
Vacuum Adaptor with Ground Joint Cone NS 45/40 lateral 2-step hose connector
  • Made of PTFE.
  • Pass-through with O-ring made of FKM for safe assembly of standard filter funnels made of glass or plastic on vessels or round bottom flasks with ground joint.
  • Lateral 2-step hose connector made of PTFE for connection to a vacuum pump by means of suitable tubing.
Specifications
Autoclavable Autoclavable
Connectors 22 mm
Disposable Reusable
Sterility Non-sterile
Diameter (Metric) 12 mm