Learn More
Thermo Scientific™ Vacutherm™ Oven Accessories and Options
Choose from a range of accessories and options for Vacutherm™ ovens to meet application-specific requirements.
Brand:  Thermo Scientific™ 51900073
124.47 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10517611
  • 161.00€ / Each
Estimated Shipment: 31-07-2024
Description
Choose from a range of Thermo Scientific™ Vacutherm™ Oven Accessories and Options to meet application-specific requirements.
Specifications
Specifications
20 hour timer (24 hr at 50Hz)
VT 6060 M/P and VT6130 M/P
Digital Timer