Learn More
Integra Biosciences™ Vacusip Aspiration System
Product Code. 11747312
Change view
Click to view available options
For Use With (Application):
Aspiration of small volumes of liquids
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
11747312 Aspiration of small volumes of liquids Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11747312 Supplier Integra Biosciences™ Supplier No. 159010
  • ONLINE EXCLUSIVE PRICE
    790.00€
    728.11€ / Pack of 1
    Save 61.89€
    7% Off
Estimated Shipment: 10-12-2026
Designed for convenient and affordable disposal of supernatant or excess liquids. Integra Biosciences™ Vacusip Aspiration System is a compact and ready-to-use vacuum aspiration system with an integrated silent vacuum pump for the aspiration of small volumes of liquids.
  • Eliminates tedious handling and the risk of contaminating house vacuum system
  • The VACUSIP is an out of the box solution
  • No installation or additional equipment is required for immediate use
  • The integrated pump runs silently and stops automatically when the vacuum is established
  • Changing a filled collection bottle of VACUSIP is clean and convenient
  • VACUSIP is small, compact and fits on every bench.
  • Due to its independence from an external vacuum source or power supply, it can be relocated wherever needed
  • When working in a safety cabinet, only a minimum of tubing and cable is desirable
  • Due to the rechargeable battery in VACUSIP, a power supply cable is not needed
  • Hydrophobic filter protects the unit from damage and contamination by aerosols and liquids.
  • For easy decontamination, all parts that come in contact with liquids can be autoclaved
Applications include aspiration and removal of:
  • Small volumes of liquid out of microcentrifuge tubes or microtiter plates.
  • Supernatants after centrifugation steps (e.g. RNA/DNA extraction)
  • Washing solutions in Western blots and ELISA
  • Remaining quantities of reagents
  • Buffers of Western blot strips
  • Excess liquid from object slides and Petri dishes
  • Bacterial solutions
  • Cell staining solutions
Specifications
For Use With (Application) Aspiration of small volumes of liquids