Learn More
Integra Biosciences™ VACUSAFE Aspiration System
Artikelnummer. 10396852
Ansicht ändern
Click to view available options
Flow Rate:
8 L/min.
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Flow Rate Packungsgröße
10396852 8 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10396852 Lieferant Integra Biosciences™ Lieferanten-Nr. 158300
  • ONLINE EXCLUSIVE PRICE
    3430.00€
    2242.97€ / Pack of 1
    Save 1187.03€
    34% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 15-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Use this aspiration system for the safe disposal of biological liquids. Integra Biosciences™ VACUSAFE Aspiration System is a compact, all-in-one laboratory vacuum pump, providing a safe and efficient way to collect and contain biological liquid waste.
  • Enables Biosafety level 1, 2 and 3 labs to comply with the stringent user safety guidelines for handling contaminated solutions
  • Maximizes productivity and minimizing noise and operational inconveniences
  • Equipped with vacuum source protection, a shatterproof bottle and a secure stand that holds the bottle in place
  • Includes integrated vacuum pump, complete with collection bottle, bottle handle, tubing set with filter, mains adapter and VACUBOY hand operator (155 510).
  • 3 liter bottle (safety coated glass)
  • Level detection and lid with quick tubing couplings (PVDF)
  • Self-closing connectors to avoid escape of drops or aerosols
  • All-in-one system includes all necessary components
  • Straightforward operation – simply switch on, set the desired vacuum and start working
  • Integrated pump / valve is switched off automatically when the vacuum is established in the bottle
  • Noise of the integrated pump is well below 65 dB(A)
  • Quick tubing connectors speed up bottle removal
Spezifikation
Includes 2 L Glass bottle, Lid with quick couplings and level detection, Tubing set with filter, Mains adapter: 100-240VAC, 50/60Hz, VACUBOY hand operator with 1-channel SS adapter 40 mm
Description VacuSafe Comfort is designed for aspiration, disposal, or filtration of biological /chemical fluids in a clean and professional way.
Flow Rate 8 L/min.
Maximum Pressure -300 to -600 mbar (adjustable)