Learn More
Cytiva VacuCap™ 90 Vacuum Filtration Devices
Product Code. 17134371
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.45 μm
1 μm
Diameter (Metric):
60 mm
90 mm
Unit Size:
Pack of 10
Product Code. 17134371
Brand: Cytiva 4622
Only null left
Add to basket
Edge
Add to basket
Innovative bottle-top filters for fast vacuum filtration of 100 mL to 5 L of aqueous solutions.
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Supor (Hydrophilic PES)
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 60 cm2
Diameter (Metric) 90 mm