Learn More
Brady™ V4500 Barcode Scanner USB Cable
Product Code. p-200022866
Click to view available options
Length (English):
3.28 ft.
9.19 ft.
Unit Size:
Each
Product Code. 30057745
Brand: Brady™ 177238
Only null left
Add to basket
Add to basket
V4500 Barcode Scanner accessories to ensure uninterrupted productivity.
Cable - USB Cable for V4500 Barcode Scanner
Specifications
Description USB to RJ50-28 Cable for V4500 Barcode Reader
Weight (Metric) 0.13 kg
Weight (English) 0.287 lb.
For Use With (Application) Asset Tracking, General Identification, Barcode Labelling, Circuit Board Labelling, Component and Equipment Labelling, Food and Beverage Process Labelling, Inventory and Inspection Labelling
For Use With (Equipment) V1300 Cradle (177235), V1300 Cradle Kit (177232), V4500 Barcode Scanner Kit (177229)
Length (English) 9.19 ft.
Type Cable
Alternate Part Number BRA-USB-RJ50-28