Learn More
BUCHI V-300 Vacuum Pump
Product Code. 15511280 Shop All Buchi Products
Change view
Click to view available options
Flow Rate:
1.8 m³/h
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
15511280 1.8 m³/h Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15511280 Supplier BUCHI Supplier No. 11V300000
Only null left
Add to basket
Add to basket
Use this pump in with the I-300 or I-300 Pro Interface for economical, speed-controlled operation and low noise emission. BUCHI V-300 Vacuum Pumps feature chemically-resistant PTFE construction and a transparent front panel. Optimally suited to work with the Rotavapor R-300.
  • Chemically resistant PTFE diaphragm pump
  • Seamless plug & play integration with the I-300/I-300 Pro Interface and the fully automated Rotavapor R-300 System
  • Contaminants are easily visible through transparent front panel
  • Unattended operation in combination with Interface I-300/I-300 Pro
  • Optional level sensor for secondary condenser alerts potential overflow of the receiving flask
  • High chemical resistance due to the use of high quality materials
  • Quiet operation and increased lifetime thanks to speed-controlled operation in conjunction with the optional Interface I-300/I-300 Pro
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 5 mbar (±2 mbar)
Includes Vacuum tubing, power cable, silencer, tubing connection set, mini-DIN communication cable, Membrane tool (to fasten and remove membrane)
Motor Speed 1500 rpm max.
Certifications/Compliance CE / CSA / CUS
Electrical Requirements 100/240 V 50/60 Hz
Description BUCHI V-300 Vacuum Pump, Stand-alone
Depth (Metric) 321 mm
Material PTFE
Model 11V300000
Noise Level 32 to 57 dBA (depending on operating mode); 32 dBA: with 10% load (normal); 57 dBA: with 100% load
Wattage 60 to 180 w
Weight (Metric) 7.6 kg
Flow Rate 1.8 m³/h
Frequency 50/60 Hz
Overall Height (Metric) 291 mm
Overall Width (Metric) 200 mm
Pump Type Oilless Diaphragm Pump
Applications General laboratory vacuum generation
Voltage 100/240 V