Learn More
UVP Replacement UV Tubes
Product Code. 12862989
Click to view available options
Wavelength:
254 nm
302 nm
365 nm
Unit Size:
Each
Product Code. 12862989
Brand: UVP 34000601
For use with 8w UV lamps.
For use with 8w UV lamps.
  • For use with 8w UV lamps
Specifications
Wattage 8 W
Wavelength 365 nm
For Use With (Equipment) Model TLW-20 transilluminator, model CL-1000 crosslinker, models 2UV and 3UV transilluminator and EL series UV lamp
Length (English) 11.33 in.
Length (Metric) 28.7 cm
Product Type Replacement UV Tube