Learn More
UVP Replacement UV Tubes
Product Code. 12822989
Change view
Click to view available options
Wavelength:
254 nm
302 nm
365 nm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Unit Size
12822989 254 nm Each
12862989 365 nm Each
11798231 302 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 12822989 Supplier UVP Supplier No. 34000701
  • ONLINE EXCLUSIVE PRICE
    51.30€
    37.85€ / Pack of 1
    Save 13.45€
    26% Off
Estimated Shipment: 21-12-2026
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
For use with 8w UV lamps.
For use with 8w UV lamps.
  • For use with 8w UV lamps
Specifications
Wattage 8 W
Wavelength 254 nm
For Use With (Equipment) CL-1000 crosslinker, DE-50 eprom eraser, models 2UV and 3UV transilluminator and EL series UV lamp
Length (English) 11.33 in.
Length (Metric) 28.7 cm
Product Type Replacement UV Tube