Learn More
UVP Replacement UV Light Tubes
Click to view available options
Wavelength:
302 nm
365 nm
Wattage:
6 W
15 W
20 W
40 W
Unit Size:
Each
Specifications
Specifications
| Wattage | 20 W |
| Wavelength | 365 nm |
| For Use With (Equipment) | Model XX-20BLB UV bench lamp |
| Length (English) | 23 in. |
| Length (Metric) | 584.2 mm |
| Packaging Quantity | 1 Ea. |
| Product Type | Replacement UV Light Tube |