Learn More
UVP J-Series Photovoltaic UV Intensity Meters: Model J-225
Product Code. 12813039
Change view
Click to view available options
Wavelength:
254nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength unitSize
12813039 254nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12813039 Supplier UVP Supplier No. 97000401
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Photovoltaic J-Series UV meters for taking accurate and repeatable readings of UV light sources in either the 365nm or 254nm UV range
  • With two scales for low and high intensity readings
  • Measures the intensity of shortwave UV (254nm). Thw sensor is sensitive within a range from 220 to 280nm with maximum sensitivity at 254nm.
  • For remote readings, a four foot (1.2m) cord connects the sensor to the meter
  • Two scales measure a wide range of intensity
  • An infrared filter assures accurate readings when measuring light sources producing infrared radiation
  • The 5× attenuation screen can be used for very high intensity lamp measurements
  • Meters are self-powered for maximum portability
All meters and sensors are individually serialized for traceability of calibration. Calibration is recommended every six months and is certified to meet UVP's published standards and is traceable to NIST. Calibrations are performed at UVP's factory.
Quality control
Specifications
Type Photovoltaic UV Intensity Meter
For Use With (Application) Accurate and repeatable readings of shortwave UV
Wavelength 254nm