Learn More
Uvex™ Cleaning Fluid
Product Code. 15396272 Shop All Uvex Products
Change view
Click to view available options
Quantity:
0.5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15396272 0.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15396272 Supplier Uvex™ Supplier No. 9972100
  • ONLINE EXCLUSIVE PRICE
    10.80€
    9.70€ / Pack of 1
    Save 1.10€
    10% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Cleaning fluid for eyeglass cleaning station
  • 0.5 L cleaning fluid for uvex™ eyeglass cleaning station (9970.002)
  • Suitable for all goggles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Lens Cleaning Solution
    For Use With (Application) Goggle cleaning
    Quantity 0.5 L
    Disposable Reusable