Learn More
Eppendorf™ UVette™
Product Code. 10412121 Shop All Eppendorf Products
Change view
Click to view available options
Quantity:
80 Pack
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10412121 80 Pack Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10412121 Supplier Eppendorf™ Supplier No. 0030106300
  • ONLINE EXCLUSIVE PRICE
    73.35€
    68.20€ / Pack of 80
    Save 5.15€
    7% Off
In Stock
Add to basket
Edge
Add to basket
Use these cuvettes for measuring small volumes (≥50μL). The Eppendorf™ UVette™ is a disposable UV/Vis cuvette with a light transmission of 220 to 1,600nm. UVettes offer superb purity, convenience and precision for photometric analysis.
  • UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
  • Two built-in optical path lengths in a single cuvette: just turn the UVette 90° to change from 10mm and 2mm
  • Volume markings for 500 and 1,000μL
  • Individually packaged and certified PCR clean and protein-free quality for sensitive (e.g. RNA) and valuable samples
  • Deep-lying optical window prevents scratches
  • Tapered cuvette base for optimal filling, frosted grip for labeling
  • Ideal for use in the Eppendorf BioPhotometer and Eppendorf BioSpectrometer, adapters available for use with other common spectrophotometers
  • Routine pack for convenient access to each UVette and safe storage in a re-closable box
TRUSTED_SUSTAINABILITY
Specifications
Type Cuvette
Capacity (Metric) 50 to 2000 μL
Material UV-transparent Plastic
Quantity 80 Pack
Path Length 2 mm or 10 mm
Wavelength Range 220 to 1600 nm
For Use With (Equipment) Biophotometer, Spectrophotometer