missing translation for 'onlineSavingsMsg'
Learn More

Vilbert Lourmat™ UV/White Light Tables

Product Code. 11880400
Click to view available options
Wavelength:
254 nm
312 nm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11880400

Brand: Vilbert Lourmat™ 215122011

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
  • 1 table, 2 functions
  • 6 tubes 8 W
  • Reading of BET (Brunauer–Emmett–Teller) gels, autoradiography, reading of protein gels

Specifications

Product Type UV/White Light Tables
Intensity 7000 mW/cm2
Wavelength 254 nm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.