Learn More
Vilber Lourmat UV Protection Face Shield
Description
- UV radiation
- Spectral curves of tubes
- 254nm = UV-C
- High bactericidal power transmitted only through quartz
- 312nm = UV-B
- 365nm = UV-A
- BLB version called Black Light or Wood Light, black tube without visible light
- User protection Imperative regardless of the wavelength emitted
- UV protection face shield that fully protects the face
- For work on light table
Specifications
Specifications
| Description | UV Protection Face Shield |
| Product Type | UV-protection faceshield |
| Model | MP-80 UV protection mask |