Learn More
Biochrom UV Grade Silica Semi-micro Cuvette with Lid and Black Walls
Click to view available options
Capacity (Metric):
400 μL
800 μL
Unit Size:
Each
Pack of 8
Description
- UV Grade Silica construction
- 10 mm semi-micro cell with lid and black walls
- 10 mm pathlength
- 4 mm internal width
- For use with Ultrospec 7000/8000/9000, Ultrospec 2100 pro, Novaspec III/Plus, GeneQuant 1300/100
Specifications
Specifications
| Type | Semi-Micro Standard Cuvette |
| Capacity (Metric) | 800 μL |
| Material | UV Grade Silica |
| Width (Metric) Interior | 4 mm |
| Quantity | 1 Ea. |
| Path Length | 10 mm |
| Wavelength Range | 190 to 1100 nm |
| Height (Metric) | 45 mm |
| For Use With (Equipment) | Spectrophotometer, Photometer |
| Width (Metric) | 12.5 mm |
| Show More |