Learn More
Benchmark Scientific™ UV-C Indicator Stickers
Click to view available options
Temperature Range (Metric):
Up to 55°C
Unit Size:
Pack of 40
Description
- Changes color in the presence of UV-C
- Used to verify operation of UV-C lamps
- 0.5 x 0.5 in. sticker
- For research use only
Specifications
Specifications
| Disposable | Single-use |
| For Use With (Application) | Confirm Operation of UV-C Lights |
| Length (Metric) | 1.25 cm |
| Product Type | UV-C Indicator Sticker |
| Max. Temperature (English) | 131°F |
| Max. Temperature (Metric) | 55°C |
| Temperature Range (English) | Up to 131°F |
| Temperature Range (Metric) | Up to 55°C |
| Calibrated | Yes |
| Display | Visual Indicator |
| Show More |